Learn More
Perkin Elmer US LLC Calcium (Ca) Pure Standard, 1,000 µg/mL, 2% HNO3, 500 mL
Catalog No. 509045648
Encompass_Preferred
Each Single Element Calibration Standard for AA and ICP-OES is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
Catalog No. 50-904-5648 Supplier Perkin Elmer US LLC Supplier No. N9300108
Add to Cart
Add to Cart
Each Single Element Calibration Standard for AA and ICP-OES is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.