Learn More
Perkin Elmer US LLC Initial Calibration Verification Standard, Matrix per Volume 5% HNO3 per 500mL
Catalog No. 509045742
Encompass_Preferred
Contents: 500 μg/mL: Ca, K, Mg, Na, 200 μg/mL: Al, Ba, 100 μg/mL: Fe, 60 μg/mL: Sb, 50 μg/mL: Co, V, 40 μg/mL: Ni, 25 μg/mL: Cu, 20 μg/mL: Zn, 15 μg/mL: Mn, 10 μg/mL: Ag, As, Cr, Tl, 5 μg/mL: Be, Cd, Se, 3 μg/mL: Pb
Catalog No. 50-904-5742 Supplier Perkin Elmer US LLC Supplier No. N9300224
Contents: 500 μg/mL: Ca, K, Mg, Na, 200 μg/mL: Al, Ba, 100 μg/mL: Fe, 60 μg/mL: Sb, 50 μg/mL: Co, V, 40 μg/mL: Ni, 25 μg/mL: Cu, 20 μg/mL: Zn, 15 μg/mL: Mn, 10 μg/mL: Ag, As, Cr, Tl, 5 μg/mL: Be, Cd, Se, 3 μg/mL: Pb