Learn More
Perkin Elmer US LLC Control Laboratory Program Instrument Calibration Solution 1 , 5% HNO3, 125 mL
Catalog No. 509045738
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 5% HNO3 5,000 µg/mL: Ca, K, Mg, Na
Catalog No. 50-904-5738 Supplier Perkin Elmer US LLC Supplier No. N9300218
Concentration of Element includes: Matrix Concentration 5% HNO3 5,000 µg/mL: Ca, K, Mg, Na