Learn More
Perkin Elmer US LLC Control Laboratory Program Instrument Calibration Solution 4 , 5% HNO3, 125 mL
Catalog No. 509045741
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 5% HNO3 100 µg/mL: As, Tl 50 µg/mL: Cd, Se, Pb
Catalog No. 50-904-5741 Supplier Perkin Elmer US LLC Supplier No. N9300221
Concentration of Element includes: Matrix Concentration 5% HNO3 100 µg/mL: As, Tl 50 µg/mL: Cd, Se, Pb