Learn More
Perkin Elmer US LLC Interference A Solution, 5% HNO3, 500 mL
Supplier:  Perkin Elmer US LLC N9300226
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 5% HNO3 5,000 µg/mL: Al, Ca, Mg 2,000 µg/mL: Fe
Catalog No. 50-904-5744