Learn More
Perkin Elmer US LLC Lead (Pb) Pure Standard, 1,000 µg/mL, 2% HNO3, 500 mL
Catalog No. 509045668
Encompass_Preferred
Each Single Element Calibration Standard for AA and ICP-OES is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
This item is not returnable. View return policy
Catalog No. 50-904-5668 Supplier Perkin Elmer US LLC Supplier No. N9300128
Add to Cart
Add to Cart
Each Single Element Calibration Standard for AA and ICP-OES is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.