Learn More
Perkin Elmer US LLC Mercury (Hg) Pure Standard, 1,000 µg/mL , 10% HNO3, 500 mL
Catalog No. 509045673
Encompass_Preferred
Each Single Element Calibration Standard for AA and ICP-OES is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
Catalog No. 50-904-5673 Supplier Perkin Elmer US LLC Supplier No. N9300133
Add to Cart
Add to Cart
Each Single Element Calibration Standard for AA and ICP-OES is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.