Learn More
Perkin Elmer US LLC Quality Control Standard 7A Elements, 5% HNO3/trace Tartaric Acid/trace HF, 100 mL
Catalog No. 509045760
Encompass_Preferred
Details Concentration Element 1,000 µg/mL K 500 µg/mL Si 100 µg/mL Al, Ba, Na 50 µg/mL Ag
Catalog No. 50-904-5760 Supplier Perkin Elmer US LLC Supplier No. N9300280
Details Concentration Element 1,000 µg/mL K 500 µg/mL Si 100 µg/mL Al, Ba, Na 50 µg/mL Ag