Learn More
Electron Microscopy Sciences Standard Carbon Adhesive Tabs, 6 mm
SDP
Catalog No. 5025109
Encompass_Preferred
Two sides of thick conductive adhesive (45 um) with the center conductive core film (35 um). Use for SEM including GSR. 125 um thick. No gas out-gassing. Traces of Si, Sb, S, Fe, Mg, Na.
Diameter: 6 mm
Catalog No. 50-251-09 Supplier Electron Microscopy Sciences Supplier No. 7782506
Two sides of thick conductive adhesive (45 um) with the center conductive core film (35 um). Use for SEM including GSR. 125 um thick. No gas out-gassing. Traces of Si, Sb, S, Fe, Mg, Na.
Diameter: 6 mm