Learn More
Andwin Scientific STD CRYOMOLD 20X25X5 1200/CS
SDP
Catalog No. NC9008137
Encompass_Preferred
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 25 mm x 20 mm x 5 mm. It is sold in a case of 1200.
Catalog No. NC9008137 Supplier Andwin Scientific Supplier No. 4557CS
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 25 mm x 20 mm x 5 mm. It is sold in a case of 1200.