Learn More
MSC Vikan Wall-/Floor Washing Brush, 305 mm, Hard, White, Euro Threaded
Catalog No. 502837550
Encompass
This wall/floor washing brush is designed for hygiene-critical environments in food manufacturing, retail, and food service. Its hard, thick bristles effectively scrub stubborn soils like fatty deposits, greases, dried batter, and biofilms from tiled floors. Compatible with all Euro threaded handles.
  • Designed for food manufacturing, retail, and food service where hygiene is critical.
  • Hard bristles effectively dislodge stubborn soils like fatty deposits, greases, dried batter, or biofilms.
  • Angled bristles reach into corners and crevices.
  • Durable construction for long-lasting daily performance.
  • Withstands harsh chemicals and cleaning agents.
  • Compatible with all Euro threaded handles for secure attachment.
  • Color-coded for hygienic zoning and 5S lean programs.
  • Easy to clean and maintain for optimal hygiene.
  • FDA compliant.
Catalog No. 50-283-7550 Supplier MSC Supplier No. 47176300
This wall/floor washing brush is designed for hygiene-critical environments in food manufacturing, retail, and food service. Its hard, thick bristles effectively scrub stubborn soils like fatty deposits, greases, dried batter, and biofilms from tiled floors. Compatible with all Euro threaded handles.
  • Designed for food manufacturing, retail, and food service where hygiene is critical.
  • Hard bristles effectively dislodge stubborn soils like fatty deposits, greases, dried batter, or biofilms.
  • Angled bristles reach into corners and crevices.
  • Durable construction for long-lasting daily performance.
  • Withstands harsh chemicals and cleaning agents.
  • Compatible with all Euro threaded handles for secure attachment.
  • Color-coded for hygienic zoning and 5S lean programs.
  • Easy to clean and maintain for optimal hygiene.
  • FDA compliant.