Learn More
Separation Systems Inc STD DHA/PIANO GASOLINE STD.
Catalog No. NC2381712
Encompass
Standard,DHA/PIANO Gasoline Standard,D6729/D6730, 6 x 1ml,
Catalog No. NC2381712 Supplier Separation Systems Inc Supplier No. SD053006
Standard,DHA/PIANO Gasoline Standard,D6729/D6730, 6 x 1ml,