Learn More
DC Scientific Glass Inc STD FIA COLUMN
SDP
Catalog No. NC9661041
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
FIA STANDARD COLUMN,Petroleum glassware,D1319 Hydrocarbon Types In Liquid Petroleum Product By Fluorescent Indicator Adsorption
Catalog No. NC9661041 Supplier DC Scientific Glass Inc Supplier No. DCP1319017
This item has a minimum qty of 2 per supplier requirements.
FIA STANDARD COLUMN,Petroleum glassware,D1319 Hydrocarbon Types In Liquid Petroleum Product By Fluorescent Indicator Adsorption