Learn More
Perkin Elmer US LLC Indium (In) Pure Standard, 1,000 µg/mL, 2% HNO3, 125 mL
Catalog No. 509047941
Encompass_Preferred
Each Single Element Calibration Standard for AA and ICP-OES is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
Catalog No. 50-904-7941 Supplier Perkin Elmer US LLC Supplier No. N9303777
Each Single Element Calibration Standard for AA and ICP-OES is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.