Learn More
Perkin Elmer US LLC Instrument Calibration Standard 3, 5% HNO3, 125 mL
Catalog No. 509047970
Encompass_Preferred
Concentration of Element includes: Concentration Content 1,000 µg/mL Ca, Fe, K, Mg, Na
Catalog No. 50-904-7970 Supplier Perkin Elmer US LLC Supplier No. N9303818
Concentration of Element includes: Concentration Content 1,000 µg/mL Ca, Fe, K, Mg, Na