Learn More
Perkin Elmer US LLC Instrument Calibration Standard 1, 5% HNO3
Catalog No. NC1533109
Encompass_Preferred
Concentration of Element includes: Concentration Content 20 µg/mL Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Ni, Pb, Sb, Se, Th, Tl, U, V, Zn
Catalog No. NC1533109 Supplier Perkin Elmer US LLC Supplier No. N9303816
Concentration of Element includes: Concentration Content 20 µg/mL Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Ni, Pb, Sb, Se, Th, Tl, U, V, Zn