Learn More
Perkin Elmer US LLC Interference Check Solution 2, 2% HNO3, 125 mL
Catalog No. 509047982
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 2% HNO3 10 µg/mL: Ag, As, Cd, Co, Cr, Cu, Mn, Ni, Zn
Catalog No. 50-904-7982 Supplier Perkin Elmer US LLC Supplier No. N9303830
Concentration of Element includes: Matrix Concentration 2% HNO3 10 µg/mL: Ag, As, Cd, Co, Cr, Cu, Mn, Ni, Zn