Learn More
Perkin Elmer US LLC Iron (Fe) Pure Standard, 1,000 µg/mL, 2% HNO3, 125 mL
Catalog No. 509046028
Encompass_Preferred
Each Single Element Calibration Standard for AA and ICP-OES is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
Catalog No. 50-904-6028 Supplier Perkin Elmer US LLC Supplier No. N9303771
Each Single Element Calibration Standard for AA and ICP-OES is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.