Learn More
ENCAPSULA NANAOSCIENCE LLC STD MACROPHAGE DEPLETN KIT 2ML
Catalog No. NC0302518
Encompass
Standard macrophage depletion kit- The kit is composed of one 2 ml vial of Clodrosome (Clodronate liposomes) and one 2 ml vial of Encapsome (plain liposomes for control).
Catalog No. NC0302518 Supplier ENCAPSULA NANAOSCIENCE LLC Supplier No. CLD89012ML
Add to Cart
Add to Cart
Standard macrophage depletion kit- The kit is composed of one 2 ml vial of Clodrosome (Clodronate liposomes) and one 2 ml vial of Encapsome (plain liposomes for control).