Learn More
Perkin Elmer US LLC ICP-MS Internal 1,2% HNO3, 125 mL
Catalog No. 509047986
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 2% HNO3 10 µg/mL: Bi, Ho, Li6, Sc, Tb, Y
Catalog No. 50-904-7986 Supplier Perkin Elmer US LLC Supplier No. N9303834
Concentration of Element includes: Matrix Concentration 2% HNO3 10 µg/mL: Bi, Ho, Li6, Sc, Tb, Y