Learn More
Bellco Glass, Inc. STD Over Head Drive Adapter,100mm
SDP
Catalog No. 501946193
Encompass_Preferred
The Bellco 100 mm Overhead Drive Cap Assembly is manufactured from 316L SST with dual bearings and seals. A dust shield for paddle assemblies (1/4” shaft) is included. Cap assembly features two 1/4” NPT female ports for addition of optional fittings. . MADE IN THE USA
Catalog No. 50-194-6193 Supplier Bellco Glass, Inc. Supplier No. 776470000
The Bellco 100 mm Overhead Drive Cap Assembly is manufactured from 316L SST with dual bearings and seals. A dust shield for paddle assemblies (1/4” shaft) is included. Cap assembly features two 1/4” NPT female ports for addition of optional fittings. . MADE IN THE USA