Learn More
Tosoh Bioscience LLC STD PEO SE-15 150000MW 0.5G
Supplier:  Tosoh Bioscience LLC 0006214
Encompass
Calibration Standard, Polyethylene Oxide; Tosoh; Description: SE-15; Molecular Weight: 1.5 x 105 M. W.; Quantity: 0.5g
Catalog No. 50-851-144