Learn More
Tosoh Bioscience LLC STD PEO SE-70 590000MW 0.5G
Supplier:  Tosoh Bioscience LLC 0006216
Encompass
Calibration Standard, Polyethylene Oxide; Tosoh; Description: SE-70; Molecular Weight: 5.9 x 105 M. W.; Quantity: 0.5g
Catalog No. 50-851-146