Learn More
Cole-Parmer Extech 601500 Standard pH Electrode, Glass; 12 x 160 mm
Supplier:  Cole-Parmer 5302642
Encompass
Replace pH electrodes when they can no longer hold calibration, dry out and cannot be rehydrated, or if the housing cracks.
Catalog No. NC1454812