Learn More
Tosoh Bioscience LLC STD PS F-1 10000MW 5G
Catalog No. 50851111 Shop All Tosoh Bioscience LLC Products
Encompass
Calibration Standard, Polystyrene; Tosoh; Description: F-1; Molecular Weight: 1.0 x 104 M. W.; Quantity: 5g
Catalog No. 50-851-111 Supplier Tosoh Bioscience LLC Supplier No. 0005207
Calibration Standard, Polystyrene; Tosoh; Description: F-1; Molecular Weight: 1.0 x 104 M. W.; Quantity: 5g