Learn More
Tosoh Bioscience LLC STD PS F-2 17000MW 5G
Supplier:  Tosoh Bioscience LLC 0005208
Encompass
Calibration Standard, Polystyrene; Tosoh; Description: F-2; Molecular Weight: 1.7 x 104 M. W.; Quantity: 5g
Catalog No. 50-851-112