Learn More
Tosoh Bioscience LLC STD PS F-380 3800000 1G
Supplier:  Tosoh Bioscience LLC 0005216
Encompass
Calibration Standard, Polystyrene; Tosoh; Description: F-380; Molecular Weight: 3.8 x 106 M. W.; Quantity: 1g
Catalog No. 50-851-120