Learn More
Tosoh Bioscience LLC STD PS F-40 420000MW 5G
Supplier:  Tosoh Bioscience LLC 0005212
Encompass
Calibration Standard, Polystyrene; Tosoh; Description: F-40; Molecular Weight: 4.2 x 105 M. W.; Quantity: 5g
Catalog No. 50-851-116