Learn More
Tosoh Bioscience LLC STD PS F-700 6800000 1G
Supplier:  Tosoh Bioscience LLC 0005219
Encompass
Calibration Standard, Polystyrene; Tosoh; Description: F-700; Molecular Weight: 6.8 x 106 M. W.; Quantity: 1g
Catalog No. 50-851-123