Learn More
Tosoh Bioscience LLC STD PS F-850 8400000MW 1G
Supplier:  Tosoh Bioscience LLC 0005220
Encompass
Calibration Standard, Polystyrene; Tosoh; Description: F-850; Molecular Weight: 8.4 x 106 M. W.; Quantity: 1g
Catalog No. 50-851-124