Learn More
Perkin Elmer US LLC Standard PVC Two-Stop Peristaltic Pump Tubing - 0.76 mm I.D., Black/Black, Pkg. 12
Catalog No. NC9350405
Encompass_Preferred
This non-flared PVC Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.
Catalog No. NC9350405 Supplier Perkin Elmer US LLC Supplier No. 09908587
This non-flared PVC Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.