Learn More
Starna Cells Inc STD RECT CELL W/STPR 1MMQ/CD
SDP
Catalog No. NC9556895
Encompass_Preferred
Standard Rectangular Quartz CD Cell with Stopper, 1mm Light Path
Catalog No. NC9556895 Supplier Starna Cells Inc Supplier No. 21Q1/CD
Standard Rectangular Quartz CD Cell with Stopper, 1mm Light Path