Learn More
MSC Ricca Chemical Company Buffer, Reference Standard, pH 10.00, Blue, 500 mL
Catalog No. 502838228
Encompass
This product is a reference standard buffer solution with a precisely calibrated pH. It is designed for accurate pH calibration and measurement in laboratory and industrial settings, ensuring reliable and consistent results.
  • Precisely calibrated to pH 10.00 ± 0.01 at 25°C
  • Color coded blue for easy identification
  • Packaged in a 500 mL natural poly container
  • 18 months shelf life
  • Suitable as a reference standard for ASTM methods for routine or continuous measurement of pH
  • Stable under normal conditions of use and storage
Catalog No. 50-283-8228 Supplier MSC Supplier No. 86393675
This product is a reference standard buffer solution with a precisely calibrated pH. It is designed for accurate pH calibration and measurement in laboratory and industrial settings, ensuring reliable and consistent results.
  • Precisely calibrated to pH 10.00 ± 0.01 at 25°C
  • Color coded blue for easy identification
  • Packaged in a 500 mL natural poly container
  • 18 months shelf life
  • Suitable as a reference standard for ASTM methods for routine or continuous measurement of pH
  • Stable under normal conditions of use and storage