Learn More
Perkin Elmer US LLC Interference Check Standards, 5% H2O/trace HNO3/0.6% Tartaric Acid, 500 mL
Catalog No. NC1756442
Encompass_Preferred
Concentration of Element includes: Matrix Concentration H2O/trace HNO306.% Tartaric Acid 1,000 µg/mL: Sb
Catalog No. NC1756442 Supplier Perkin Elmer US LLC Supplier No. N9303797
Add to Cart
Add to Cart
Concentration of Element includes: Matrix Concentration H2O/trace HNO306.% Tartaric Acid 1,000 µg/mL: Sb