Learn More
ERA WATERS CORP STD SET FOR SIMPLE TOC P & A
SDP
Catalog No. NC1862962
Encompass
"Contains one each of #18001 Reagent Water (Rw) and #18002 0.500 ppb USP Sucrose (Rs) - 40 mLThis CRM is produced under ISO 9001, ISO/IEC 17025 and ISO 17034 (previously Guide 34) accredited quality system."
Catalog No. NC1862962 Supplier ERA WATERS CORP Supplier No. 18009
"Contains one each of #18001 Reagent Water (Rw) and #18002 0.500 ppb USP Sucrose (Rs) - 40 mLThis CRM is produced under ISO 9001, ISO/IEC 17025 and ISO 17034 (previously Guide 34) accredited quality system."