Learn More
MSC Proto Standard Socket, 1/4 in Drive, 6 Point, 1 ea
Catalog No. 501920306
Encompass
Proto sockets with TorquePlus are designed for use on stubborn or damaged fasteners, as well as fasteners made from soft materials. They feature a full polish and meet aerospace specification AS4984A, ensuring a thicker nickel-chrome coating for enhanced corrosion resistance and durability.
  • Standard socket type
  • Drive size is 1/4 in
  • Socket size is 1/4 in
  • Features 6 points
  • Made of alloy steel
  • Has a chrome finish
  • Overall length is 7/8 in
  • Not tether capable
  • Meets AS4984A and ASME B107.110-2012 standards
Catalog No. 50-192-0306 Supplier MSC Supplier No. 39940390
Proto sockets with TorquePlus are designed for use on stubborn or damaged fasteners, as well as fasteners made from soft materials. They feature a full polish and meet aerospace specification AS4984A, ensuring a thicker nickel-chrome coating for enhanced corrosion resistance and durability.
  • Standard socket type
  • Drive size is 1/4 in
  • Socket size is 1/4 in
  • Features 6 points
  • Made of alloy steel
  • Has a chrome finish
  • Overall length is 7/8 in
  • Not tether capable
  • Meets AS4984A and ASME B107.110-2012 standards