Learn More
MSC Proto Hand Socket, 3/8 in, 12.00 mm, 6-Point, 1 ea
Catalog No. 501920378
Encompass
The Proto Standard Hand Socket features a 3/8 in drive and a 12.00 mm socket with a 6-point design. Made from durable alloy steel, this standard length socket boasts a chrome finish and is not tether capable. It conforms to AS4984A and ASME B107.110-2012 standards and is manufactured in the United States.
  • Standard socket type
  • Features a 12.00 mm socket size
  • Has a 3/8 in drive size
  • Six points
  • Overall length of 28.6000 decimal inches
  • Made from alloy steel
  • Standard length type
  • Not tether capable
  • Conforms to AS4984A and ASME B107.110-2012 standards
  • Features a chrome finish
  • Manufactured in the United States
Catalog No. 50-192-0378 Supplier MSC Supplier No. 39936687
The Proto Standard Hand Socket features a 3/8 in drive and a 12.00 mm socket with a 6-point design. Made from durable alloy steel, this standard length socket boasts a chrome finish and is not tether capable. It conforms to AS4984A and ASME B107.110-2012 standards and is manufactured in the United States.
  • Standard socket type
  • Features a 12.00 mm socket size
  • Has a 3/8 in drive size
  • Six points
  • Overall length of 28.6000 decimal inches
  • Made from alloy steel
  • Standard length type
  • Not tether capable
  • Conforms to AS4984A and ASME B107.110-2012 standards
  • Features a chrome finish
  • Manufactured in the United States