Learn More
MSC Proto Standard Socket, 3/8 in. Drive, 13/16, 12-Point, 1 ea
Catalog No. 501920395
Encompass
This standard hand socket is designed for industrial and professional applications, providing reliable performance for fastening and loosening tasks. Manufactured from durable alloy steel, it features a chrome finish for enhanced corrosion resistance.
  • Standard socket type for general purpose use
  • Socket size of 13/16 in. for specific fastener applications
  • Drive size of 3/8 in. compatible with standard ratchets and extensions
  • Features a 12-point configuration for secure grip and engagement
  • Constructed from alloy steel for strength and durability
  • Chrome finish provides resistance to corrosion
  • Meets AS4984A and ASME B107.110-2012 standards
Catalog No. 50-192-0395 Supplier MSC Supplier No. 39932884
This standard hand socket is designed for industrial and professional applications, providing reliable performance for fastening and loosening tasks. Manufactured from durable alloy steel, it features a chrome finish for enhanced corrosion resistance.
  • Standard socket type for general purpose use
  • Socket size of 13/16 in. for specific fastener applications
  • Drive size of 3/8 in. compatible with standard ratchets and extensions
  • Features a 12-point configuration for secure grip and engagement
  • Constructed from alloy steel for strength and durability
  • Chrome finish provides resistance to corrosion
  • Meets AS4984A and ASME B107.110-2012 standards