Learn More
MSC Proto Socket, 3/8 in Drive, 15/16 in, 12-Point, 1 ea
Catalog No. 501920397
Encompass
Proto standard hand sockets are designed for efficient fastening and loosening of nuts and bolts. This 3/8 in drive, 15/16 in 12-point socket is made from durable alloy steel with a chrome finish.
  • Standard socket type
  • Socket size: 15/16 in
  • Drive size: 3/8 in
  • Features 12 points for secure grip
  • Overall length measures 1-5/16 in
  • Made from alloy steel
  • Chrome finish for durability
  • Meets AS4984A and ASME B107.110-2012 standards
  • Not tether capable
Catalog No. 50-192-0397 Supplier MSC Supplier No. 39932645
Proto standard hand sockets are designed for efficient fastening and loosening of nuts and bolts. This 3/8 in drive, 15/16 in 12-point socket is made from durable alloy steel with a chrome finish.
  • Standard socket type
  • Socket size: 15/16 in
  • Drive size: 3/8 in
  • Features 12 points for secure grip
  • Overall length measures 1-5/16 in
  • Made from alloy steel
  • Chrome finish for durability
  • Meets AS4984A and ASME B107.110-2012 standards
  • Not tether capable