Learn More
MSC Proto Socket, 3/8 in Drive, 16 mm, 12-Point, 1 ea
Catalog No. 501920372
Encompass
This standard hand socket is engineered for reliable performance in various fastening applications. It features a durable chrome finish and specialized lobular openings designed for efficient engagement and greater turning power.
  • Chrome finish
  • Designed for quick engagement on both 6 and 12-point fasteners
  • Lobular openings for greater turning power without deforming the fastener
Catalog No. 50-192-0372 Supplier MSC Supplier No. 39934419
This standard hand socket is engineered for reliable performance in various fastening applications. It features a durable chrome finish and specialized lobular openings designed for efficient engagement and greater turning power.
  • Chrome finish
  • Designed for quick engagement on both 6 and 12-point fasteners
  • Lobular openings for greater turning power without deforming the fastener