Learn More
MSC Proto Hand Socket, 3/8 in Drive, 16 mm, 6 Point
Catalog No. 501920382
Encompass
Proto 6-point sockets are engineered for challenging applications, effectively handling stubborn or damaged fasteners, as well as those made from soft materials. Featuring the TorquePlus™ design, these sockets offer quick and easy engagement to fastener flats, shifting contact stresses away from corners for enhanced performance.
  • High-performance drive configuration
  • Quick and easy socket engagement to fastener flats
  • Minimizes slippage and fastener rounding
  • Extends fastener life
  • Allows user to more easily turn worn, rounded, or rusted fasteners
Catalog No. 50-192-0382 Supplier MSC Supplier No. 39936802
Proto 6-point sockets are engineered for challenging applications, effectively handling stubborn or damaged fasteners, as well as those made from soft materials. Featuring the TorquePlus™ design, these sockets offer quick and easy engagement to fastener flats, shifting contact stresses away from corners for enhanced performance.
  • High-performance drive configuration
  • Quick and easy socket engagement to fastener flats
  • Minimizes slippage and fastener rounding
  • Extends fastener life
  • Allows user to more easily turn worn, rounded, or rusted fasteners