Learn More
MSC Proto Hand Socket, 3/8 in, 3/4 in, 12-Point, 1 ea
Catalog No. 501920394
Encompass
A standard hand socket designed for general-purpose fastening, featuring a 3/8 in drive, 3/4 in socket size, and a 12-point configuration for secure grip.
  • Overall length of 1-3/16 in
  • Chrome finish for durability and corrosion resistance
  • Constructed from alloy steel
  • Meets AS4984A and ASME B107.110-2012 standards
Catalog No. 50-192-0394 Supplier MSC Supplier No. 39934989
A standard hand socket designed for general-purpose fastening, featuring a 3/8 in drive, 3/4 in socket size, and a 12-point configuration for secure grip.
  • Overall length of 1-3/16 in
  • Chrome finish for durability and corrosion resistance
  • Constructed from alloy steel
  • Meets AS4984A and ASME B107.110-2012 standards