Learn More
MSC Proto Standard Hand Socket, 3/8 in, 6-Point, Chrome, 1 ea
Catalog No. 501920359
Encompass
This standard hand socket is designed for high performance and durability, featuring a chrome finish for corrosion resistance. Its TorquePlus™ design makes it ideal for use on stubborn or damaged fasteners, as well as those made from soft materials.
  • Standard socket type
  • A 3/8 inch drive size
  • A 3/8 inch socket size
  • A 6-point configuration
  • Chrome finish for durability and corrosion resistance
  • TorquePlus™ design for use on stubborn or damaged fasteners
  • Suitable for fasteners made from soft materials
Catalog No. 50-192-0359 Supplier MSC Supplier No. 39934484
This standard hand socket is designed for high performance and durability, featuring a chrome finish for corrosion resistance. Its TorquePlus™ design makes it ideal for use on stubborn or damaged fasteners, as well as those made from soft materials.
  • Standard socket type
  • A 3/8 inch drive size
  • A 3/8 inch socket size
  • A 6-point configuration
  • Chrome finish for durability and corrosion resistance
  • TorquePlus™ design for use on stubborn or damaged fasteners
  • Suitable for fasteners made from soft materials