Learn More
MSC Proto Standard Socket, 1/4 in Drive, 5/16, 12-Point, 1 ea
Catalog No. 501920328
Encompass
The Proto J4710T is a 1/4 in drive, 5/16 in, 12-point standard hand socket crafted from alloy steel with a full polished chrome finish. It features a TorquePlus design, enabling effective engagement with fasteners, including those that are worn, rounded, or made from soft materials.
  • Radius-corner socket design engages flats for better grip
  • Improves the ability to turn worn or rounded fasteners
  • Minimizes fastener rounding and helps extend fastener life
  • TorquePlus design is ideal for use on stubborn or damaged fasteners
  • Distributes contact stresses over a large area of the socket, minimizing slippage
  • Full polished chrome finish provides greater corrosion resistance and durability
  • Meets and exceeds stringent aerospace specification AS4984A
Catalog No. 50-192-0328 Supplier MSC Supplier No. 39932553
The Proto J4710T is a 1/4 in drive, 5/16 in, 12-point standard hand socket crafted from alloy steel with a full polished chrome finish. It features a TorquePlus design, enabling effective engagement with fasteners, including those that are worn, rounded, or made from soft materials.
  • Radius-corner socket design engages flats for better grip
  • Improves the ability to turn worn or rounded fasteners
  • Minimizes fastener rounding and helps extend fastener life
  • TorquePlus design is ideal for use on stubborn or damaged fasteners
  • Distributes contact stresses over a large area of the socket, minimizing slippage
  • Full polished chrome finish provides greater corrosion resistance and durability
  • Meets and exceeds stringent aerospace specification AS4984A