Learn More
MSC Proto Socket, 3/8 in, 12 Point, 5/8 in, 1 ea
Catalog No. 501920392
Encompass
This standard socket features a radius-corner design for superior grip, making it ideal for stubborn, worn, or damaged fasteners, as well as those made from soft materials. Its full polish and thick nickel-chrome coating ensure enhanced corrosion resistance and durability, meeting stringent aerospace specifications.
  • Radius-corner socket design
  • Engages flats for better grip
  • Improves ability to turn worn or rounded fasteners
  • Ideal for stubborn or damaged fasteners
  • Suitable for fasteners made from soft materials
  • Full polish for corrosion resistance
  • Thicker nickel-chrome coating for greater durability
  • Meets and exceeds stringent aerospace specification AS4984A
Catalog No. 50-192-0392 Supplier MSC Supplier No. 39935085
This standard socket features a radius-corner design for superior grip, making it ideal for stubborn, worn, or damaged fasteners, as well as those made from soft materials. Its full polish and thick nickel-chrome coating ensure enhanced corrosion resistance and durability, meeting stringent aerospace specifications.
  • Radius-corner socket design
  • Engages flats for better grip
  • Improves ability to turn worn or rounded fasteners
  • Ideal for stubborn or damaged fasteners
  • Suitable for fasteners made from soft materials
  • Full polish for corrosion resistance
  • Thicker nickel-chrome coating for greater durability
  • Meets and exceeds stringent aerospace specification AS4984A