Learn More
MSC Proto Standard Socket, 1/4 in, 6 mm, 6-Point, 1 ea
Catalog No. 501920318
Encompass
This standard hand socket is designed for general-purpose use, featuring a 1/4 in drive and a 6 mm, 6-point configuration. It is finished with chrome for durability and corrosion resistance.
  • Alloy steel construction for strength
  • Six-point socket design for secure fastener engagement
  • Overall length of 7/8 in for standard applications
  • Chrome finish for enhanced corrosion resistance
  • Meets AS4984A and ASME B107.110-2012 standards
  • TorquePlus design minimizes fastener rounding and extends fastener life
Catalog No. 50-192-0318 Supplier MSC Supplier No. 39940705
This standard hand socket is designed for general-purpose use, featuring a 1/4 in drive and a 6 mm, 6-point configuration. It is finished with chrome for durability and corrosion resistance.
  • Alloy steel construction for strength
  • Six-point socket design for secure fastener engagement
  • Overall length of 7/8 in for standard applications
  • Chrome finish for enhanced corrosion resistance
  • Meets AS4984A and ASME B107.110-2012 standards
  • TorquePlus design minimizes fastener rounding and extends fastener life