Learn More
MSC Proto Socket, 3/8 in. Drive, 7/16 in., 12-Point, 1 ea
Catalog No. 501920389
Encompass
Proto sockets are engineered for professional use, featuring a TorquePlus™ design that effectively handles stubborn, damaged, or soft material fasteners. This design minimizes slippage and fastener rounding, ensuring enhanced durability and easier operation.
  • Ideal for stubborn, damaged, or soft material fasteners
  • TorquePlus™ design for quick, easy engagement to fastener flats, not corners
  • Radius-corner design shifts stress away from fastener corners, minimizing slippage
  • Internal TorquePlus™ design minimizes fastener rounding and extends fastener life
Catalog No. 50-192-0389 Supplier MSC Supplier No. 39932611
Proto sockets are engineered for professional use, featuring a TorquePlus™ design that effectively handles stubborn, damaged, or soft material fasteners. This design minimizes slippage and fastener rounding, ensuring enhanced durability and easier operation.
  • Ideal for stubborn, damaged, or soft material fasteners
  • TorquePlus™ design for quick, easy engagement to fastener flats, not corners
  • Radius-corner design shifts stress away from fastener corners, minimizing slippage
  • Internal TorquePlus™ design minimizes fastener rounding and extends fastener life