Learn More
Grainger Std Stl Blk 2.5 -12 3.75x2.209 Grd8 1PK
Catalog No. 502752346
Encompass_Preferred
The Grainger 14A835 is a 14 gauge, type TFN/THHN, 600 volt insulated copper wire for use in residential and commercial buildings. It is UL and CSA listed for installation in conduit, cable trays, ducts, plenums, and other approved raceways. This general purpose building wire is available in multiple colors and 1000 foot spools. It can be used for branch circuits, feeders, and services requiring high temperature and moisture resistance.
Catalog No. 50-275-2346 Supplier Grainger Supplier No. 14A835
The Grainger 14A835 is a 14 gauge, type TFN/THHN, 600 volt insulated copper wire for use in residential and commercial buildings. It is UL and CSA listed for installation in conduit, cable trays, ducts, plenums, and other approved raceways. This general purpose building wire is available in multiple colors and 1000 foot spools. It can be used for branch circuits, feeders, and services requiring high temperature and moisture resistance.