Learn More
Ace Glass, Inc. Adapter, maxi, Ace-Thred, with 7506-06 bushing, 24/40 inner to#15 Ace-Thred
SDP
Catalog No. NC1827528
Encompass_Preferred
Adapter with ground joint at bottom and either a #11, #15, #25 or #36 Ace-Thred at top.
Catalog No. NC1827528 Supplier Ace Glass, Inc. Supplier No. 503040
Adapter with ground joint at bottom and either a #11, #15, #25 or #36 Ace-Thred at top.