Learn More
Chemglass Life Sciences Chemglass Std Wall Lab Tubing Size 8, Excelon PVC, 1/4 in ID x 1/2 in OD x 50 ft (CG-730-008)
SDP
Catalog No. NC2754845
Encompass_Preferred
Standard-wall PVC laboratory tubing for general fluid transfer and vacuum applications. Size 8 tubing measures 1/4 in ID x 1/2 in OD with a 1/8 in wall and is supplied on 50 ft rolls. The tubing provides flexible, smooth-walled construction suitable for fittings and clamps used in laboratory setups.
  • PVC construction with a smooth interior for consistent flow.
  • 1/4 in inner diameter, 1/2 in outer diameter, and 1/8 in wall thickness.
  • Supplied as a 50 ft roll for extended runs.
  • Suitable for general fluid transfer and vacuum applications.
  • Flexible and compatible with standard laboratory barbed fittings and clamps.
Catalog No. NC2754845 Supplier Chemglass Life Sciences Supplier No. CG730008
Standard-wall PVC laboratory tubing for general fluid transfer and vacuum applications. Size 8 tubing measures 1/4 in ID x 1/2 in OD with a 1/8 in wall and is supplied on 50 ft rolls. The tubing provides flexible, smooth-walled construction suitable for fittings and clamps used in laboratory setups.
  • PVC construction with a smooth interior for consistent flow.
  • 1/4 in inner diameter, 1/2 in outer diameter, and 1/8 in wall thickness.
  • Supplied as a 50 ft roll for extended runs.
  • Suitable for general fluid transfer and vacuum applications.
  • Flexible and compatible with standard laboratory barbed fittings and clamps.